Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFPL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112687
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112687 Supplier Invitrogen™ Supplier No. PA5112687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFPL3 Polyclonal Antibody for Western Blot, ELISA

RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFPL3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RFPL3
Gene Accession No. O75679
Gene Alias ret finger protein like 3; ret finger protein-like 3; RFPL3
Gene Symbols RFPL3
Host Species Rabbit
Immunogen Recombinant Human Ret finger protein-like 3 protein (8-288AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10738
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.