Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFPL4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531938 Supplier Invitrogen™ Supplier No. PA531938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RFPL4B gene ontology (GO) annotation includes metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFPL4B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RFPL4B
Gene Accession No. Q6ZWI9
Gene Alias ret finger protein like 4B; ret finger protein-like 4B; RFPL4B; RING finger protein 211; RNF211
Gene Symbols RFPL4B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RFPL4B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.