Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537032 Supplier Invitrogen™ Supplier No. PA537032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 63 kDa. Purity is >95% by SDS-PAGE.

Raftlin, also known as RFTN1 (raftlin, lipid raft linker 1), cell migration-inducing gene 2 protein, PIB10, PIG9 or MIG2, is a 578 amino acid cell membrane protein and lipid anchor that is essential for raft cell assembly and maintenance. A member of the raftlin family, Raftlin modulates B-cell antigen receptor-mediated signaling, TCR signals, and is involved in T cell-mediated immune responses. The gene encoding Raftlin maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFTN1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Rftn1
Gene Accession No. Q14699, Q6A0D4
Gene Alias 2310015n21rik; AI853526; Cell migration-inducing gene 2 protein; KIAA0084; MIG2; PIB10; PIG9; proliferation-inducing protein 10; proliferation-inducing protein 9; Raftlin; raftlin lipid raft linker 1; raftlin, lipid raft linker 1; raft-linking protein; Rftn1; RGD1310777; RGD1563347
Gene Symbols Rftn1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 475-520 of human Raftlin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23180, 501095, 76438
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.