Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFTN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557202 Supplier Invitrogen™ Supplier No. PA557202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVAAKRGMKF VGFISQHYSP SKFCNGTNHD GDIESMLHVR HGSDENCRSW NEGTLSGQSS ESGIEEELHH ESGQYQMEQN GSPTSSKSRK GE Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 58%.

Upon bacterial lipopolysaccharide stimulation, mediates clathrin-dependent internalization of TLR4 in dendritic cells, resulting in activation of TICAM1-mediated signaling and subsequent IFNB1 production. May regulate B-cell antigen receptor-mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFTN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFTN2
Gene Accession No. Q52LD8
Gene Alias 2700010E02Rik; 3222401M22Rik; C2orf11; raftlin family member 2; raftlin-2; Raft-linking protein 2; RFTN2; RGD1307304
Gene Symbols RFTN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFTN2. Recombinant protein control fragment (Product #RP-96170).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.