Learn More
RFWD3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301398A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Pig, Orangutan, Monkey, Gorilla and Chimpanzee
E3 ubiquitin-protein ligase that mediates the ubiquitination of p53/TP53 in the late response to DNA damage, and acts as a positive regulator of p53/TP53 stability, thereby regulating the G1/S DNA damage checkpoint. May act by catalyzing the formation of short polyubiquitin chains on p53/TP53 that are not targeted to the proteasome. In response to ionizing radiation, interacts with MDM2 and enhances p53/TP53 ubiquitination, possibly by restricting MDM2 from extending polyubiquitin chains on ubiquitinated p53/TP53. Plays a role in RPA-mediated DNA damage signaling and repair. {ECO:0000269; PubMed:20173098, ECO:0000269; PubMed:21504906, ECO:0000269; PubMed:21558276}.Specifications
| RFWD3 | |
| Polyclonal | |
| Unconjugated | |
| RFWD3 | |
| RNF201, E3 ubiquitin-protein ligase RFWD3, RING finger and WD repeat domain-containing protein 3, RING finger protein 201 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55159 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q6PCD5 | |
| RFWD3 | |
| Between 724 and 774 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.