Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFX1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB183791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183791 Supplier Novus Biologicals Supplier No. NBP282031AF700
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RFX1 Polyclonal antibody specifically detects RFX1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen RFX1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias EFC, EF-C, Enhancer factor C, MHC class II regulatory factor RFX, MHC class II regulatory factor RFX1, Regulatory factor X 1, regulatory factor X, 1 (influences HLA class II expression), RFX, trans-acting regulatory factor 1, Transcription factor RFX1
Host Species Rabbit
Immunogen RFX1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RFX1. The immunogen is located within the last 50 amino acids of RFX1. Amino Acid Squence: AGGESPALGPETLEPPAK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5989
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.