Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590912 Supplier Invitrogen™ Supplier No. PA590912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQVHSPPEQS PVQANSSSSK TAGAPTGTVP QQLQVHGVQQ SVPVTQERSV VQATPQAPKP GPVQPLTVQG LQPVHVAQEV QQLQQVPVPH VYSSQVQYVE G; Positive Samples: LO2; Cellular Location: Nucleus.

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X2, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with RFX family members X2, X3, and X5, but not with X4. This protein binds to the X-boxes of MHC class II genes and is essential for their expression. Also, it can bind to an inverted repeat that is required for expression of hepatitis B virus genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RFX1
Gene Accession No. P22670
Gene Alias AI047719; AI385641; EFC; EF-C; Enhancer factor C; MHC class II regulatory factor RFX; MHC class II regulatory factor RFX1; Regulatory factor X 1; regulatory factor X, 1 (influences HLA class II expression); regulatory factor X1; RFX; RFX1; trans-acting regulatory factor 1; transcription factor RFX1; vregulatory factor X, 1 (influences HLA class II expression)
Gene Symbols RFX1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human RFX1 (NP_0029094).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5989
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.