Learn More
Invitrogen™ RFX2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total RFX2 protein.
Specifications
Specifications
| Antigen | RFX2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RFX2 |
| Gene Accession No. | P48378 |
| Gene Alias | DNA binding protein RFX2; DNA-binding protein RFX2; FLJ14226; HLA class II regulatory factor RFX2; regulatory factor X 2; regulatory factor X, 2 (influences HLA class II expression); regulatory factor X2; RFX2; trans-acting regulatory factor 2 |
| Gene Symbols | RFX2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.