Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFX3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP131658

Catalog No. NBP131658


Only null left
Add to Cart

Description

Description

RFX3 Polyclonal specifically detects RFX3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RFX3
Polyclonal
Unconjugated
1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.01% Thimerosal
RFX3
Recombinant protein encompassing a sequence within the center region of human RFX3. The exact sequence is proprietary.
Affinity Purified
RUO
5991
Human
IgG
Western Blot
0.43mg/mL
Western Blot 1:500-1:3000
bA32F11.1, DNA binding protein RFX3, MGC87155, Regulatory factor X 3, regulatory factor X, 3 (influences HLA class II expression), transcription factor RFX3
Rabbit
84 kDa
0.1 mL
Primary
Tested in human. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only