Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFX3 Antibody (OTI3F7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273865AF405
Description
RFX3 Monoclonal specifically detects RFX3 in Human, Mouse samples. It is validated for Western Blot.Specifications
RFX3 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RFX3 | |
Full length recombinant protein of human RFX3 (NP_602304) produced in HEK293T cells. | |
0.1 mL | |
5991.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3F7 | |
Western Blot | |
bA32F11.1, DNA binding protein RFX3, MGC87155, Regulatory factor X 3, regulatory factor X, 3 (influences HLA class II expression), transcription factor RFX3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction