Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFX3 Antibody (OTI3F7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273865
Description
RFX3 Monoclonal specifically detects RFX3 in Human, Mouse samples. It is validated for Western Blot.Specifications
RFX3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
bA32F11.1, DNA binding protein RFX3, MGC87155, Regulatory factor X 3, regulatory factor X, 3 (influences HLA class II expression), transcription factor RFX3 | |
Mouse | |
83.3 kDa | |
100 μg | |
5991 | |
Human, Mouse | |
Lyophilized |
Western Blot | |
OTI3F7 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RFX3 | |
Full length human recombinant protein of human RFX3 (NP_602304) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction