Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527805

Catalog No. PIPA527805


Only null left
Add to Cart

Description

Description

Recommended positive controls: A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (83%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. Two transcript variants encoding different isoforms have been described for this gene, and at least one of the variants utilizes alternative polyadenylation signals.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RFX3
Polyclonal
Unconjugated
RFX3
bA32F11.1; C230093O12Rik; DNA binding protein RFX3; MRFX3; Regulatory factor X 3; regulatory factor X, 3 (influences HLA class II expression); regulatory factor X3; Rfx2; RFX3; Transcription factor RFX3
Rabbit
Antigen affinity chromatography
RUO
5991
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.3 mg/mL
PBS with 1% BSA, 10% glycerol and 0.01% thimerosal; pH 7
P48380
RFX3
Recombinant protein encompassing a sequence within the center region of human RFX3. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.