Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFX4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24896725UL
Description
RFX4 Polyclonal antibody specifically detects RFX4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
RFX4 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
NYD-SP10, Regulatory factor X 4, regulatory factor X, 4 (influences HLA class II expression), regulatory factor X4, Testis development protein NYD-SP10, transcription factor NYD-sp10, transcription factor RFX4 | |
This antibody was developed against a recombinant protein corresponding to amino acids: STTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPH | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5992 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction