Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562205 Supplier Invitrogen™ Supplier No. PA562205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STTGAMQSYT WSLTYTVTTA AGSPAENSQQ LPCMRNTHVP SSSVTHRIPV YPHREEHGYT GSYNYGSYGN QHPHPMQSQY PALPH Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X3, and X5. It has been shown to interact with itself as well as with regulatory factors X2 and X3, but it does not interact with regulatory factor X1. This protein may be a transcriptional repressor rather than a transcriptional activator. Three transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFX4
Gene Accession No. Q33E94
Gene Alias 4933412G19Rik; NYD-sp10; Regulatory factor X 4; regulatory factor X 4 protein; regulatory factor X, 4 (influences HLA class II expression); regulatory factor X4; RFX4; RGD1562092; Testis development protein NYD-SP10; transcription factor RFX4; winged-helix transcription factor RFX4
Gene Symbols RFX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFX4. Recombinant protein control fragment (Product #RP-101198).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.