Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFX5 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100195JF646
Description
RFX5 Polyclonal antibody specifically detects RFX5 in Human samples. It is validated for Western BlotSpecifications
RFX5 | |
Polyclonal | |
Western Blot | |
DNA-binding protein RFX5, Regulatory factor X 5, regulatory factor X, 5 (influences HLA class II expression) | |
Rabbits were immunized with a synthetic peptide, which represented a portion of human Regulatory Factor X 5 encoded within exon 5 (LocusLink ID 5993), conjugated to KLH. After multiple immunizations in Freund's adjuvant, serum was collected. Antibodies were affinity purified using the peptide immobilized on solid support. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
5993 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction