Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558065 Supplier Invitrogen™ Supplier No. PA558065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHCSTYPEPI YPTLPQANHD FYSTSSNYQT VFRAQPHSTS GLYPHHTEHG RCMAWTEQQL SRDFFSGSCA GSPYNSRPPS SYGPSLQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

RFX6 is a member of the regulatory factor X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX6
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFX6
Gene Accession No. Q8HWS3
Gene Alias 4930572O07Rik; dJ955L16.1; DNA-binding protein RFX6; MTCHRS; MTFS; Regulatory factor X 6; regulatory factor X domain containing 1; regulatory factor X domain-containing protein 1; regulatory factor X, 6; regulatory factor X6; RFX6; RFXDC1; si:dkeyp-93d12.2
Gene Symbols RFX6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFX6. Recombinant protein control fragment (Product #RP-95023).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.