Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568655 Supplier Invitrogen™ Supplier No. PA568655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

RFX6 is a member of the regulatory factor X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RFX6
Gene Accession No. Q8C7R7
Gene Alias 4930572O07Rik; dJ955L16.1; DNA-binding protein RFX6; MTCHRS; MTFS; Regulatory factor X 6; regulatory factor X domain containing 1; regulatory factor X domain-containing protein 1; regulatory factor X, 6; regulatory factor X6; RFX6; RFXDC1; si:dkeyp-93d12.2
Gene Symbols RFX6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse RFX6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320995
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.