Learn More
RFX7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303061A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RFX7 is a member of the regulatory factor X (RFX) family of transcription factors.Specifications
RFX7 | |
Polyclonal | |
Unconjugated | |
RFX7 | |
RFXDC2, DNA-binding protein RFX7, regulatory factor X domain containing 2, regulatory factor X domain-containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64864 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q2KHR2 | |
RFX7 | |
Between 400 and 450 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.