Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFX8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546693 Supplier Invitrogen™ Supplier No. PA546693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AIINQGTLAT SKKALASDRS GADELENNPE MKCLRNLISL LGTSTDLRVF Sequence homology: Human: 100%.

RFX8 may be a transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RFX8
Gene Accession No. Q6ZV50
Gene Alias DNA-binding protein RFX8; Regulatory factor X 8; regulatory factor X, 8; RFX family member 8, lacking RFX DNA binding domain; RFX gene family member 8, lacking RFX DNA binding domain; RFX8
Gene Symbols RFX8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RFX8 (aa 321-370).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 731220
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.