Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFXANK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112212 Supplier Invitrogen™ Supplier No. PA5112212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RFXANK Polyclonal Antibody for Western Blot, IP

Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. This protein contains ankyrin repeats involved in protein-protein interactions. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group B. Two transcript variants encoding different isoforms have been described for this gene, with only one isoform showing activation activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFXANK
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene RFXANK
Gene Accession No. O14593
Gene Alias ANKRA1; Ankyrin repeat family A protein 1; ankyrin repeat-containing regulatory factor X-associated protein; BLS; DNA-binding protein RFXANK; F14150_1; regulatory factor X associated ankyrin containing protein; regulatory factor X subunit B; Regulatory factor X-associated ankyrin-containing protein; RFXANK; RFXB; RFX-B; RFX-Bdelta4
Gene Symbols RFXANK
Host Species Rabbit
Immunogen E. coli-derived Human RFXANK fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.