Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFXAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565115 Supplier Invitrogen™ Supplier No. PA565115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVKLEESADNI LSIVKQRTGS FGDRPARPTL LEQVLNQKRL SLLRSPEVVQ FLQKQQQLLN QQVLEQRQQQ FPGT Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated ankyrin-containing protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group D. Transcript variants utilizing different polyA signals have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFXAP
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFXAP
Gene Accession No. O00287
Gene Alias 5730495K23Rik; regulatory factor X associated protein; regulatory factor X-associated protein; RFX DNA-binding complex 36 kDa subunit; RFXAP; RFX-AP; RFX-associated protein; si:ch211-170h2.4
Gene Symbols RFXAP
Host Species Rabbit
Immunogen Recombinant Human RFXAP. Recombinant protein control fragment (Product #RP-105877).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.