Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RFXAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568883
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated ankyrin-containing protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group D. Transcript variants utilizing different polyA signals have been found for this gene.
Specifications
RFXAP | |
Polyclonal | |
Unconjugated | |
RFXAP | |
5730495K23Rik; regulatory factor X associated protein; regulatory factor X-associated protein; RFX DNA-binding complex 36 kDa subunit; RFXAP; RFX-AP; RFX-associated protein; si:ch211-170h2.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170767 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8VCG9 | |
RFXAP | |
synthetic peptide corresponding to a region of mouse RFXAP (aa 101-150). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction