Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ RG 108
GREENER_CHOICESDP

Catalog No. 329550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-955-0 50 mg
32-951-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-955-0 Supplier R&D Systems™ Supplier No. 3295/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-nucleoside DNA methyltransferase inhibitor

Non-nucleoside DNA methyltransferase inhibitor that blocks the enzyme active site. Inhibits DNA methylation in human cancer cell lines in vitro without detectable toxicity. Demethylates and reactivates epigenetically silenced tumor suppressor genes. Enhances the efficiency of induced pluripotent stem cell (iPS) generation.

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 334.33
Purity >99%
Molecular Weight (g/mol) 334.331
Chemical Name or Material RG 108
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.