Learn More
Invitrogen™ RG1 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
$459.00 - $1159.00
Specifications
Antigen | RG1 Biosimilar |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
Classification | Recombinant Monoclonal |
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term 19G9.
RG1 is a gene that has primarily been studied in the context of Ginsenoside Rg1, a bioactive compound derived from the roots of Panax ginseng. Ginsenoside Rg1 is known for its neuroprotective effects and regulatory role in immune responses. It modulates various signaling pathways, including the phosphatidylinositol 3-kinase (PI3K)/Akt pathway, which is crucial for cell survival and growth. Additionally, Rg1 has been shown to inhibit apoptotic pathways mediated by endoplasmic reticulum stress and the c-Jun N-terminal kinase (JNK) pathway in neurodegenerative disease models, potentially offering therapeutic benefits in conditions like Alzheimer's disease. Furthermore, Rg1 influences the activation of astrocytes through long non-coding RNAs (lncRNAs) and microRNAs, contributing to the repair of spinal cord injuries. Overall, RG1, through its association with Ginsenoside Rg1, plays a significant role in cellular protection and modulation across various biological systems.Specifications
RG1 Biosimilar | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Lyophilized | |
IgG1 | |
Human | |
Primary | |
Protein A |
1 mg/mL | |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
Unconjugated | |
Human | |
RUO | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.