Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RG9MTD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | RG9MTD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RG9MTD1 Polyclonal specifically detects RG9MTD1 in Human samples. It is validated for Western Blot.Specifications
RG9MTD1 | |
Polyclonal | |
Rabbit | |
Q7L0Y3 | |
54931 | |
Synthetic peptides corresponding to RG9MTD1(RNA (guanine-9-) methyltransferase domain containing 1) The peptide sequence was selected from the middle region of RG9MTD1. Peptide sequence KKARQIKKEMKAAAREEAKNIKLLETTEEDKQKNFLFLRLWDRNMDIAMG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.1.1, EC 2.1.1.-, EC 2.1.1.36, FLJ20432, HBV pre-S2 trans-regulated protein 2, Mitochondrial RNase P protein 1, MRPP1mitochondrial ribonuclease P protein 1, NY-REN-49 antigen, Renal carcinoma antigen NY-REN-49, RNA (guanine-9-) methyltransferase domain containing 1, RNA (guanine-9-)-methyltransferase domain-containing protein 1 | |
TRMT10C | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title