Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RG9MTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541873 Supplier Invitrogen™ Supplier No. PA541873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KKARQIKKEM KAAAREEAKN IKLLETTEED KQKNFLFLRL WDRNMDIAMG Sequence homology: Human: 100%; Rabbit: 93%.

This gene encodes the precursor of a subunit of the mitochondrial ribonuclease P, which is involved in 5' processing of mitochondrial tRNAs. The encoded protein may confer RNA-binding capacity to mitochondrial ribonuclease P and may be essential for transcript processing, RNA modification, translation and mitochondrial respiration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RG9MTD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRMT10C
Gene Accession No. Q7L0Y3
Gene Alias COXPD30; FLJ20432; HBV pre-S2 trans-regulated protein 2; HNYA; mitochondrial ribonuclease P protein 1; Mitochondrial RNase P protein 1; mitochondrial RNase P subunit 1; mRNA methyladenosine-N(1)-methyltransferase; MRPP1; Renal carcinoma antigen NY-REN-49; RG9MTD1; RNA (guanine-9-) methyltransferase domain containing 1; RNA (guanine-9-)-methyltransferase domain-containing protein 1; TRMT10C; tRNA (adenine(9)-N(1))-methyltransferase; tRNA (guanine(9)-N(1))-methyltransferase; tRNA methyltransferase 10 homolog C; tRNA methyltransferase 10C, mitochondrial RNase P subunit
Gene Symbols TRMT10C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RG9MTD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54931
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.