Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RG9MTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561552
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561552 Supplier Invitrogen™ Supplier No. PA561552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQMEPNSDGH DRKRVRRDVV HSTLRLIIDC SFDHLMVLKD IKKLHKQIQR CYAENRRALH PVQFYLTSHG GQLKKNMDEN DKGWVNW Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Probable RNA methyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RG9MTD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRMT10A
Gene Accession No. Q8TBZ6
Gene Alias epididymis secretory protein Li 88; HEL-S-88; MSSGM; MSSGM1; RG9MTD2; RNA (guanine-9-)-methyltransferase domain-containing protein 2; TRM10; TRMT10A; tRNA (guanine(9)-N(1))-methyltransferase TRMT10A; tRNA methyltransferase 10 homolog A; tRNA methyltransferase 10A
Gene Symbols TRMT10A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RG9MTD2. Recombinant protein control fragment (Product #RP-96676).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.