Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RG9MTD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528521 Supplier Invitrogen™ Supplier No. PA528521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji. Predicted reactivity: Mouse (87%), Rat (86%), Rhesus Monkey (97%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

S-adenosyl-L-methionine-dependent guanine N(1)-methyltransferase that catalyzes the formation of N(1)-methylguanine at position 9 (m1G9) in tRNAs (PubMed:23042678). Probably not able to catalyze formation of N(1)-methyladenine at position 9 (m1A9) in tRNAs (PubMed:23042678). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RG9MTD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TRMT10B
Gene Accession No. Q6PF06
Gene Alias 2610042J10Rik; bA3J10.9; RG9MTD3; RNA (guanine-9-) methyltransferase domain containing 3; RNA (guanine-9-)-methyltransferase domain-containing protein 3; Rnmtd3; RP11-3J10.9; Trmt10b; tRNA (guanine(9)-N(1))-methyltransferase TRMT10B; tRNA methyltransferase 10 homolog B; tRNA methyltransferase 10 homolog B (S. cerevisiae); tRNA methyltransferase 10B
Gene Symbols TRMT10B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RG9MTD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158234
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.