Learn More
RGAG1 Polyclonal Antibody

Description
RGAG1 Polyclonal Antibody for IHC
Specifications
Specifications
| Antigen | RGAG1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RTL9 |
| Gene Accession No. | Q8NET4 |
| Gene Alias | Gm385; Kiaa1318; Mar9; Mart9; retrotransposon gag domain containing 1; retrotransposon gag domain-containing protein 1; Retrotransposon Gag-like protein 9; Rgag1; RTL9; Sushi-XF2; tumor antigen BJHCC23; tumor antigen BJ-HCC-23 |
| Gene Symbols | RTL9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.