Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101206 Supplier Invitrogen™ Supplier No. PA5101206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RGAG1.

RTL9 is one of at least 11 genes called Mar or Mart related to long terminal repeat retrotransposons. They do not correspond to functional retrotransposons, but rather to neofunctionalized retrotransposons genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGAG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RTL9
Gene Accession No. Q32KG4, Q8NET4
Gene Alias Gm385; Kiaa1318; Mar9; Mart9; retrotransposon gag domain containing 1; retrotransposon gag domain-containing protein 1; Retrotransposon Gag-like protein 9; Rgag1; RTL9; Sushi-XF2; tumor antigen BJHCC23; tumor antigen BJ-HCC-23
Gene Symbols RTL9
Host Species Rabbit
Immunogen A synthesized peptide derived from human RTL9(Accession Q8NET4), corresponding to amino acid residues N1339-K1388.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209540, 57529
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.