Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABCOCO1 Polyclonal Antibody
CABCOCO1 Polyclonal Antibody
Supplier: Thermo Scientific PA570132
Description
CABCOCO1 Polyclonal Antibody for Western Blot

Specifications
CABCOCO1 | |
Polyclonal | |
Unconjugated | |
CABCOCO1 | |
bA63A2.1; C10orf107; CABCOCO1; chromosome 10 open reading frame 107; Ciliary-associated calcium-binding coiled-coil protein 1; RGD1306739; similar to RIKEN cDNA 1700040L02; uncharacterized protein C10orf107; uncharacterized protein C10orf107 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
361834 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
CABCOCO1 | |
synthetic peptide directed towards the C-terminal region of rat RGD1306739. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction