Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RGFP 966
GREENER_CHOICESDP

Catalog No. 67285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-285 5 mg
67-282-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-285 Supplier Tocris Bioscience™ Supplier No. 6728/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective HDAC3 inhibitor (IC50 = 80 nM)

RGFP 966 is a potent and selective HDAC3 inhibitor (IC50 = 80 nM); exhibits no effect on other HDAC (up to 15 μM). RGFP 966 reduces the frequency of striatal CAG repeat expansions in mouse models of Huntington's disease and reduces mHTT accumulation. RGFP 966 enhances long-term object-location memory in vivo.

Specifications

Chemical Name or Material (2E)-N-(2-Amino-4-fluorophenyl)-3-[1-(3-phenyl-2-propen-1-yl)-1H-pyrazol-4-yl]-2-propenamide
CAS 1357389-11-7
Quantity 5 mg
Molecular Formula C21H19FN4O
Formula Weight 362.4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.