Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554081 Supplier Invitrogen™ Supplier No. PA554081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHFPCLQKLL DYLTRMMPGS DPERRAQNLL EQFQKQEVET DNGLPNTISF SLEEEEELEG GESAEFTCFS EDLVAEQLTY MDAQLFKKVV PHHCLGCIWS RRD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Ral GDS (Ral guanine nucleotide dissociation stimulator) is a guanine nucleotide exchange factor (GEF) that activates Ral and is implicated in oncogenic Ras-induced cell transformation. RGL1 (ral guanine nucleotide dissociation stimulator-like 1), also known as RGL or RalGDS-like 1, is a 768 amino acid protein that is a putative GEF. Strongly expressed in brain, heart, spleen, kidney and testis, RGL1 is a downstream effector protein that is involved in Ras and Ral signaling pathways. RGL1 contains an N-terminal Ras-GEF domain and a C-terminal Ras-interacting domain that interacts with the GTP-bound form of Ras through its effector loop. Due to its similarity to Ral GDS, RGL1 may be implicated in carcinogenesis. Two isoforms exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGL1
Gene Accession No. Q9NZL6
Gene Alias KIAA0959; mKIAA0959; ral guanine nucleotide dissociation stimulator like 1; ral guanine nucleotide dissociation stimulator,-like 1; ral guanine nucleotide dissociation stimulator-like 1; RalGDS-like 1; RGL; Rgl1
Gene Symbols RGL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGL1. Recombinant protein control fragment (Product #RP-92439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.