Learn More
RGL2 Mouse anti-Human, Clone: OTI10F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808391

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.Specifications
RGL2 | |
Monoclonal | |
Unconjugated | |
RGL2 | |
HKE1.5, KE1.5, RAB2L | |
Mouse | |
Affinity Chromatography | |
RUO | |
5863 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI10F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15211 | |
RGL2 | |
Human recombit protein fragment corresponding to amino acids 547-777 of human RGL2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.