Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGL2 Antibody (OTI10F2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273875JF646
Description
RGL2 Monoclonal specifically detects RGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RGL2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RGL2 | |
Recombinant protein fragment corresponding to amino acids 547-777 of human RGL2 (NP_004752) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI10F2 | |
Western Blot, Immunohistochemistry | |
GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
5863.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction