Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528280 Supplier Invitrogen™ Supplier No. PA528280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (93%), Pig (94%), Rhesus Monkey (100%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene RGL2
Gene Accession No. O15211
Gene Alias GDS-related protein; HKE1.5; KE1.5; RAB2L; ral guanine nucleotide dissociation stimulator like 2; ral guanine nucleotide dissociation stimulator,-like 2; ral guanine nucleotide dissociation stimulator-like 2; RalGDS-like 2; ralGDS-like factor; Ras-associated protein RAB2L; Rgl2; Rgt2; Rlf
Gene Symbols RGL2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGL2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.