Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583851
Description
Immunogen sequence: FDKRRKEFAV LSELRRLQNE CRGYNLQPDH DIQRWLQGLR PLTEAQSHRV SCEVEPPGSS DPPAPRVLRP TLVISQWA.
Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.
Specifications
RGL2 | |
Polyclonal | |
Unconjugated | |
RGL2 | |
GDS-related protein; HKE1.5; KE1.5; RAB2L; ral guanine nucleotide dissociation stimulator like 2; ral guanine nucleotide dissociation stimulator,-like 2; ral guanine nucleotide dissociation stimulator-like 2; RalGDS-like 2; ralGDS-like factor; Ras-associated protein RAB2L; Rgl2; Rgt2; Rlf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5863 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15211 | |
RGL2 | |
Recombinant protein corresponding to Human RGL2. Recombinant protein control fragment (Product #RP-108377). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction