Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560438
Description
Immunogen sequence: VEIKKTAVQD LSFNKNLRAV VSVLGSWLQD HPQDFRDHPA HSDLGSVRTF LGWAAPGSAE AQKAEKLLED Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Specifications
RGL3 | |
Polyclonal | |
Unconjugated | |
RGL3 | |
ral guanine nucleotide dissociation stimulator like 3; ral guanine nucleotide dissociation stimulator-like 3; RalGDS-like 3; RalGEF-like protein 3, mouse homolog; RGL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57139 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3MIN7 | |
RGL3 | |
Recombinant protein corresponding to Human RGL3. Recombinant protein control fragment (Product #RP-99234). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction