Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557492
Description
Immunogen sequence: KLLTNLPAAA VLSAQVYSAV LRGLWEENVC GTPGRTRVCT ALLYGQVCPF QDSTDGLRTI TSILFNWPPE NTSVYYQPPQ RSSFRIKLAF RNLSWPGLGL EDHQEIVLGQ LVLPEPNEAK PDDPAPRPGQ HAL Highest antigen sequence identity to the following orthologs: Mouse - 20%, Rat - 20%.

Specifications
RGL4 | |
Polyclonal | |
Unconjugated | |
RGL4 | |
hRGR; ral guanine nucleotide dissociation stimulator like 4; ral guanine nucleotide dissociation stimulator-like 4; RalGDS related oncogene; Ral-GDS related protein Rgr; ralGDS-like 4; ral-GDS-related protein; RGL4; RGR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
266747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZJ4 | |
RGL4 | |
Recombinant protein corresponding to Human RGL4. Recombinant protein control fragment (Product #RP-91551). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction