Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531507
Description
Recommended positive controls: NT2D1, U87-MG. Predicted reactivity: Human (99%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
RGL4 | |
Polyclonal | |
Unconjugated | |
RGL4 | |
hRGR; ral guanine nucleotide dissociation stimulator like 4; ral guanine nucleotide dissociation stimulator-like 4; RalGDS related oncogene; Ral-GDS related protein Rgr; ralGDS-like 4; ral-GDS-related protein; RGL4; RGR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
266747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IZJ4 | |
RGL4 | |
Recombinant protein fragment corresponding to a region within amino acids 18 and 309 of Human RGL4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction