Learn More
Invitrogen™ RGMA Monoclonal Antibody (275407)

Description
In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human RGM-A and recombinant chicken RGM is observed. In Western blots, approximately 25% cross-reactivity with recombinant mouse RGM-C is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | RGMA |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 275407 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | RGMA |
| Gene Accession No. | Q6PCX7, Q8JG54, Q96B86 |
| Gene Alias | BC059072; C230063O06; repulsive guidance molecule; repulsive guidance molecule A; repulsive guidance molecule family member a; RGM; RGM domain family member A; RGM domain family, member A; RGMA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.