Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGMB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547516
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547516 Supplier Invitrogen™ Supplier No. PA547516
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) RGM-A and rmRGM-C is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Repulsive Guidance Molecule B (RGMB) is primarily expressed in the central nervous system. RGM A and B both appear at every level of the developing neural axis, where they colocalize to a large extent in the mantle layer, although only RGMA appears in the neuroepithelium, and only RGMB in the peripheral nervous system. During development, both RGMA and B appear also in lung and in limb cartilage, while RGMB has additional expression domains in pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGMB
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene RGMB
Gene Accession No. Q7TQ33
Gene Alias 1110059F19Rik; DRAGON; DRG11-responsive axonal guidance and outgrowth of neurite; repulsive guidance molecule B; repulsive guidance molecule family member B; RGM domain family member B; RGM domain family, member B; RGMB
Gene Symbols RGMB
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RGM-B Gly49-Ser414.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68799
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.