Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGMB/Repulsive Guidance Molecule B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1396 Supplier Bioss Supplier No. BS11474R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Repulsive Guidance Molecule B (RGMB) is primarily expressed in the central nervous system. RGM A and B both appear at every level of the developing neural axis, where they colocalize to a large extent in the mantle layer, although only RGMA appears in the neuroepithelium, and only RGMB in the peripheral nervous system. During development, both RGMA and B appear also in lung and in limb cartilage, while RGMB has additional expression domains in pancreas.

Specifications

Antigen RGMB/Repulsive Guidance Molecule B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RGMB
Gene Accession No. Q6NW40
Gene Alias 1110059F19Rik; DRAGON; DRG11-responsive axonal guidance and outgrowth of neurite; repulsive guidance molecule B; repulsive guidance molecule family member B; RGM domain family member B; RGM domain family, member B; RGMB
Gene Symbols RGMB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Repulsive Guidance Molecule B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285704
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.