Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556056 Supplier Invitrogen™ Supplier No. PA556056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALYSLFPDHH VKKYFDQVDI SNGLDWSLDH KIFYYIDSLS YSVDAFDYDL QTGQISNRRS VYKLEKEEQI PDGMC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

The protein encoded by this gene is a highly conserved, calcium-binding protein, that is preferentially expressed in the liver and kidney. It may have an important role in calcium homeostasis. Studies in rat indicate that this protein may also play a role in aging, as it shows age-associated down-regulation. This gene is part of a gene cluster on chromosome Xp11.3-Xp11.23. Alternative splicing results in two transcript variants having different 5' UTRs, but encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rgn
Gene Accession No. Q15493
Gene Alias AI265316; epididymis secretory protein Li 41; gluconolactonase; GNL; HEL-S-41; RC; Reguc; regucalcin; regucalcin (senescence marker protein-30); Rgn; Senescence marker protein 30; Smp30; SMP-30
Gene Symbols Rgn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGN. Recombinant protein control fragment (Product #RP-95632).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9104
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.