Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGNEF Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222282F
Description
RGNEF Polyclonal antibody specifically detects RGNEF in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
RGNEF | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
190 kDa guanine nucleotide exchange factor, p190RHOGEF, Rho-guanine nucleotide exchange factor | |
The immunogen this antibody was made to, maps to a region between residue 1655 and 1705 of human 190 kDa, Rho-Guanine Nucleotide Exchange Factor using the numbering given in entry NP_001171164.1 (GeneID 64283). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64283 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction