Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281799MFV450
Description
RGP1 Polyclonal antibody specifically detects RGP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| RGP1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| retrograde Golgi transport protein RGP1 homolog, RGP1 retrograde golgi transport homolog (S. cerevisiae) | |
| RGP1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of the human RGP1. The immunogen is located within amino acids 50 - 100 of RGP1. Amino Acid Squence: ALPPPDSSQPDVQPDS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 9827 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction