Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520519

Catalog No. PIPA520519


Only null left
Add to Cart

Description

Description

A suggested positive control is human heart tissue lysate. PA5-20519 can be used with blocking peptide PEP-0639.

Retrograde golgi transport homolog 1 (RGP1) is the mammalian homolog to the yeast RGP1, a protein that forms a tight complex with RIC1. This complex binds Ypt6p and stimulates guanine nucleotide exchange. RGP1 is localized to the Golgi and is thought to be a potential Golgi recycling factor. Rgp1 yeast mutants exhibit defects in retrograde trafficking similar to those seen in yeast with mutations in other retrograde Golgi transport proteins. It is expected that RGP1 plays a similar role in mammalian cells to that seen in yeast.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGP1
Polyclonal
Unconjugated
RGP1
1110029E03Rik; AI527196; F730001J03; KIAA0258; mKIAA0258; RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; RGD1311757; RGP1; RGP1 (HGNC:21965); RGP1 homolog, RAB6A GEF complex partner 1; RGP1 retrograde golgi transport homolog; RGP1 retrograde golgi transport homolog (S. cerevisiae)
Rabbit
Antigen affinity chromatography
RUO
242406, 313493, 9827
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8BHT7, Q92546
RGP1
A 17 amino acid peptide near the amino terminus of the human RGP1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.