Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554325 Supplier Invitrogen™ Supplier No. PA554325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKSYSYSEVL PIEGPPSFRG QSVKYVYKLT IGCQRVNSPI TLLRVPLRVL VLTGLQDVRF PQDEAVAPSS PFLEEDEGGK KDS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Retrograde golgi transport homolog 1 (RGP1) is the mammalian homolog to the yeast RGP1, a protein that forms a tight complex with RIC1. This complex binds Ypt6p and stimulates guanine nucleotide exchange. RGP1 is localized to the Golgi and is thought to be a potential Golgi recycling factor. Rgp1 yeast mutants exhibit defects in retrograde trafficking similar to those seen in yeast with mutations in other retrograde Golgi transport proteins. It is expected that RGP1 plays a similar role in mammalian cells to that seen in yeast.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGP1
Gene Accession No. Q92546
Gene Alias 1110029E03Rik; AI527196; F730001J03; KIAA0258; mKIAA0258; RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; RGD1311757; RGP1; RGP1 (HGNC:21965); RGP1 homolog, RAB6A GEF complex partner 1; RGP1 retrograde golgi transport homolog; RGP1 retrograde golgi transport homolog (S. cerevisiae)
Gene Symbols RGP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGP1. Recombinant protein control fragment (Product #RP-92906).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.