Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGPD Pan Polyclonal Antibody
RGPD Pan Polyclonal Antibody
Supplier: Thermo Scientific PA5103247
Description
RGPD Pan Polyclonal Antibody for Western Blot

Specifications
RGPD Pan | |
Polyclonal | |
Unconjugated | |
RGPD2 | |
BS63; BS-63; epididymis luminal protein 161; HEL161; nucleoporin; NUP358; RAN binding protein 2-like 1; Ran binding protein 2-like 2; ran-binding protein 2-like 1; ran-binding protein 2-like 1/2; Ran-binding protein 2-like 2; ran-binding protein 2-like 3; RanBP2alpha; RANBP2L1; RANBP2L2; RANBP2L3; ranBP2-like 1; ranBP2-like 1/2; RanBP2-like 2; ranBP2-like 3; RANBP2-like and GRIP domain containing 2; RANBP2-like and GRIP domain containing 3; RANBP2-like and GRIP domain containing 4; RANBP2-like and GRIP domain containing 5; RANBP2-like and GRIP domain containing 8; RANBP2-like and GRIP domain-containing protein 2; RanBP2-like and GRIP domain-containing protein 3; ranBP2-like and GRIP domain-containing protein 4; RANBP2-like and GRIP domain-containing protein 5; RANBP2-like and GRIP domain-containing protein 5/6; RANBP2-like and GRIP domain-containing protein 8; RGP2; RGP3; RGP4; RGP5; RGP6; RGP7; RGP8; RGPD2; RGPD3; RGPD4; RGPD5; RGPD6; RGPD7; RGPD8; Sperm membrane protein BS-63 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
285190, 653489, 727851, 729857, 84220 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A6NKT7, O14715, P0DJD1, Q7Z3J3, Q99666 | |
RGPD2, RGPD3, RGPD4, RGPD5, RGPD8 | |
A synthesized peptide derived from human RGPD8(Accession O14715), corresponding to amino acid residues S1586-K1636. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction