Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGPD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561078 Supplier Invitrogen™ Supplier No. PA561078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APLTEAAHRH FTIEKHGDSK WIIYRFTKQL CGTERARAKI S Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 27%.

The RANBP2-like and GRIP domain containing 5 protein (RGPD5) has high similarity to RANBP2, a large RAN-binding protein localized at the cytoplasmic side of the nuclear pore complex. The gene coding for RGPD5 is thought to have arisen from a gene duplication event of RANBP2 as these highly homologous genes are located close to each other at chromosome 2q11-q12. RGPD5 was identified as an HIV dependency factor (HDF), suggesting that RGPD5 may be an important drug target in HIV treatment. At least two isoforms of RGPD5 are known to exist, of which the shorter isoform is expressed primarily in testis, while the longer of the two is expressed at low levels in a number of somatic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGPD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGPD5
Gene Accession No. Q99666
Gene Alias BS63; BS-63; epididymis luminal protein 161; HEL161; ran-binding protein 2-like 1; ran-binding protein 2-like 1/2; RANBP2L1; RANBP2L2; ranBP2-like 1; ranBP2-like 1/2; RANBP2-like and GRIP domain containing 5; RANBP2-like and GRIP domain-containing protein 5; RANBP2-like and GRIP domain-containing protein 5/6; RANBPL1; RGP5; RGP6; RGP7; RGPD5; RGPD6; RGPD7; Sperm membrane protein BS-63
Gene Symbols RGPD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGPD5. Recombinant protein control fragment (Product #RP-104397).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.